ENA375227839 Enamine

2-amino-2-phenyl-N-[(thiophen-2-yl)methyl]acetamide

  • MDL number MFCD11650853
  • Cas Number : 1218009-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115358
Availability details for
Quantity
  • Availability
  • Add to Cart