ENA375227852 Enamine

2-[3-(2-methoxyethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD18733934
  • Cas Number : 1486485-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115424
Availability details for
Quantity
  • Availability
  • Add to Cart