ENA375227855 Enamine

N-(2-aminoethyl)-1-methyl-1H-pyrrole-3-sulfonamide

  • MDL number MFCD11633199
  • Cas Number : 1096270-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115428
Availability details for
Quantity
  • Availability
  • Add to Cart