ENA375227856 Enamine

6-amino-2-(4-methoxyphenyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD12189042
  • Cas Number : 1154345-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115429
Availability details for
Quantity
  • Availability
  • Add to Cart