ENA375227857 Enamine

N-(1,3-dioxaindan-5-yl)formamide

  • MDL number MFCD21121151
  • Cas Number : 117889-14-2
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114348
Availability details for
Quantity
  • Availability
  • Add to Cart