ENA375227864 Enamine

methyl 6-chloro-2-methylpyridine-3-carboxylate

  • MDL number MFCD14155903
  • Cas Number : 851759-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30...32
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115443
Availability details for
Quantity
  • Availability
  • Add to Cart