ENA375227865 Enamine

2,2-difluoro-2-(1-hydroxycyclobutyl)acetic acid

  • MDL number MFCD16749774
  • Cas Number : 681240-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115444
Availability details for
Quantity
  • Availability
  • Add to Cart