ENA375227868 Enamine

4-bromo-6-(trifluoromethoxy)-1,3-benzothiazol-2-amine

  • MDL number MFCD22565986
  • Cas Number : 144631-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202...204
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115476

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115476
Availability details for
Quantity
  • Availability
  • Add to Cart