ENA375227873 Enamine

4-(chloromethyl)-2-(4-chlorophenyl)-6-methylpyrimidine

  • MDL number MFCD16868246
  • Cas Number : 1340094-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115483
Availability details for
Quantity
  • Availability
  • Add to Cart