ENA375227875 Enamine

2-(quinoxalin-2-ylsulfanyl)ethane-1-sulfonyl chloride

  • MDL number MFCD19637935
  • Cas Number : 1342703-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114383

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114383
Availability details for
Quantity
  • Availability
  • Add to Cart