ENA375227881 Enamine

5-(difluoromethoxy)-2-fluoropyridine

  • MDL number MFCD13185839
  • Cas Number : 130318-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115499
Availability details for
Quantity
  • Availability
  • Add to Cart