ENA375227894 Enamine

(3R,4S)-3-[(1,3-dioxaindan-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride

  • MDL number MFCD22565990
  • Cas Number : 130855-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-138
Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115519
Availability details for
Quantity
  • Availability
  • Add to Cart