ENA375227910 Enamine

2-amino-N-cyclopentyl-N-ethylpentanamide

  • MDL number MFCD12105403
  • Cas Number : 1218401-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115563
Availability details for
Quantity
  • Availability
  • Add to Cart