ENA375227915 Enamine

2-(4-tert-butylphenyl)propan-2-amine

  • MDL number MFCD11581287
  • Cas Number : 17797-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115575
Availability details for
Quantity
  • Availability
  • Add to Cart