ENA375227919 Enamine

2-amino-N-[(3-fluorophenyl)methyl]-N-methylpentanamide

  • MDL number MFCD12105297
  • Cas Number : 1218040-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115580
Availability details for
Quantity
  • Availability
  • Add to Cart