ENA375227920 Enamine

1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine

  • MDL number MFCD04970058
  • Cas Number : 1006436-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115582

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115582
Availability details for
Quantity
  • Availability
  • Add to Cart