ENA375227923 Enamine

5-(furan-2-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD22565996
  • Cas Number : 1423032-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115590

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115590
Availability details for
Quantity
  • Availability
  • Add to Cart