ENA375227927 Enamine

2-(2,4,6-trimethylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD08276815
  • Cas Number : 3167-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300...302
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115594

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115594
Availability details for
Quantity
  • Availability
  • Add to Cart