ENA375227933 Enamine

2-(4-fluoro-3-methylphenyl)-2-oxoacetaldehyde

  • MDL number MFCD08448035
  • Cas Number : 81593-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114405

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114405
Availability details for
Quantity
  • Availability
  • Add to Cart