ENA375227935 Enamine

2-(1,4-dithian-2-yl)acetic acid

  • MDL number MFCD22566003
  • Cas Number : 933690-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115628
Availability details for
Quantity
  • Availability
  • Add to Cart