ENA375227936 Enamine

5-chloro-1-methyl-1H-1,3-benzodiazole-2-carbaldehyde

  • MDL number MFCD18807802
  • Cas Number : 156212-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115629

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115629
Availability details for
Quantity
  • Availability
  • Add to Cart