ENA375227946 Enamine

1-iodonaphthalene-2-carbonitrile

  • MDL number MFCD18411476
  • Cas Number : 1261562-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115651
Availability details for
Quantity
  • Availability
  • Add to Cart