ENA375227948 Enamine

2-(pyridin-2-yl)-1-(pyridin-3-yl)ethan-1-one

  • MDL number MFCD00225313
  • Cas Number : 59576-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115658
Availability details for
Quantity
  • Availability
  • Add to Cart