ENA375227949 Enamine

N-(2-hydroxyethyl)oxane-4-sulfonamide

  • MDL number MFCD16666273
  • Cas Number : 1427967-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115659
Availability details for
Quantity
  • Availability
  • Add to Cart