ENA375227950 Enamine

(5-amino-1,3,4-thiadiazol-2-yl)methanol

  • MDL number MFCD12766834
  • Cas Number : 56951-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114425

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-114425
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart