ENA375227953 Enamine

2-(dimethyl-1,2-oxazol-4-yl)propan-1-amine

  • MDL number MFCD08701359
  • Cas Number : 893641-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115704

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115704
Availability details for
Quantity
  • Availability
  • Add to Cart