ENA375227963 Enamine

4-chloro-2,6-bis(propan-2-yl)pyrimidine

  • MDL number MFCD12137679
  • Cas Number : 1155152-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108681
Availability details for
Quantity
  • Availability
  • Add to Cart