ENA375227966 Enamine

4,6-dimethylazepan-2-one

  • MDL number MFCD19219003
  • Cas Number : 45827-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115719

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115719
Availability details for
Quantity
  • Availability
  • Add to Cart