ENA375227973 Enamine

(6-methoxypyridazin-3-yl)methanol

  • MDL number MFCD14606401
  • Cas Number : 89532-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-57
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115729
Availability details for
Quantity
  • Availability
  • Add to Cart