ENA375227976 Enamine

2-methyl-1-[3-(4H-1,2,4-triazol-3-yl)-1,2,4-oxadiazol-5-yl]propan-1-amine

  • MDL number MFCD11120072
  • Cas Number : 1042521-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115738
Availability details for
Quantity
  • Availability
  • Add to Cart