ENA375227980 Enamine

3-ethyl-2-methyl-6-phenoxy-1,4-dihydroquinolin-4-one

  • MDL number MFCD18466770
  • Cas Number : 1178442-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115744
Availability details for
Quantity
  • Availability
  • Add to Cart