ENA375227986 Enamine

ethyl 2-(5-amino-1,3,4-thiadiazol-2-yl)acetate

  • MDL number MFCD00620684
  • Cas Number : 88124-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-115755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115755
Availability details for
Quantity
  • Availability
  • Add to Cart