ENA375227989 Enamine

N-(1,2,3,4-tetrahydroquinolin-3-yl)acetamide

  • MDL number MFCD09810049
  • Cas Number : 945-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114447
Availability details for
Quantity
  • Availability
  • Add to Cart