ENA375227990 Enamine

3-amino-1,6-dimethyl-1,2-dihydropyridin-2-one

  • MDL number MFCD22566019
  • Cas Number : 1423034-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115761
Availability details for
Quantity
  • Availability
  • Add to Cart