ENA375227993 Enamine

4-ethyl-1,3-thiazol-5-amine

  • MDL number MFCD16669882
  • Cas Number : 1249955-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114453
Availability details for
Quantity
  • Availability
  • Add to Cart