ENA375228001 Enamine

1-(4-phenylquinolin-2-yl)piperidin-3-ol

  • MDL number MFCD22566024
  • Cas Number : 1423027-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115826
Availability details for
Quantity
  • Availability
  • Add to Cart