ENA375228004 Enamine

4-methoxybenzene-1-carboximidamide hydrochloride

  • MDL number MFCD00466114
  • Cas Number : 51721-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217-220
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-115842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115842
Availability details for
Quantity
  • Availability
  • Add to Cart