ENA375228013 Enamine

2-bromo-1H-indene

  • MDL number MFCD06797863
  • Cas Number : 10485-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35...37
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114511

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-114511
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart