ENA375228021 Enamine

4-(2-methoxyethoxy)benzene-1-carbothioamide

  • MDL number MFCD09942523
  • Cas Number : 1016788-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115902
Availability details for
Quantity
  • Availability
  • Add to Cart