ENA375228027 Enamine

(5-methyl-1,3-oxazol-2-yl)methyl acetate

  • MDL number MFCD11111631
  • Cas Number : 888022-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115909

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115909
Availability details for
Quantity
  • Availability
  • Add to Cart