ENA375228029 Enamine

1-chloro-4,7-difluoro-2,3-dihydro-1H-indene

  • MDL number MFCD11219579
  • Cas Number : 1188233-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115911

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115911
Availability details for
Quantity
  • Availability
  • Add to Cart