ENA375228035 Enamine

1,4,6-trichloro-2,3-dihydro-1H-indene

  • MDL number MFCD11219573
  • Cas Number : 1188146-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115918
Availability details for
Quantity
  • Availability
  • Add to Cart