ENA375228042 Enamine

N-[(2-chlorophenyl)methyl]-4-fluoroaniline

  • MDL number MFCD03210821
  • Cas Number : 356531-97-0
  • Molecular Weight : 235.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-chlorobenzyl)-4-fluoroaniline
Inchi Code 1S/C13H11ClFN/c14-13-4-2-1-3-10(13)9-16-12-7-5-11(15)6-8-12/h1-8,16H,9H2
Inchi Key DYLHNBFNWGXIQQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115931
Availability details for
Quantity
  • Availability
  • Add to Cart