ENA375228049 Enamine

1H-pyrazole-5-carbothioamide

  • MDL number MFCD05662460
  • Cas Number : 1314902-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115938
Availability details for
Quantity
  • Availability
  • Add to Cart