ENA375228052 Enamine

2-(4-fluorobenzenesulfonamido)-N'-hydroxyethanimidamide

  • Cas Number : 1251417-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114522
Availability details for
Quantity
  • Availability
  • Add to Cart