ENA375228057 Enamine

4-(4-chlorophenoxy)-3-fluorobenzonitrile

  • MDL number MFCD12110251
  • Cas Number : 1153105-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115953

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115953
Availability details for
Quantity
  • Availability
  • Add to Cart