ENA375228062 Enamine

1,4-dichloro-2,3-dihydro-1H-indene

  • MDL number MFCD11219558
  • Cas Number : 1082562-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116000
Availability details for
Quantity
  • Availability
  • Add to Cart