ENA375228068 Enamine

4-(chloromethyl)-2-(4-fluorophenyl)pyrimidine

  • MDL number MFCD16866707
  • Cas Number : 1339350-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116006
Availability details for
Quantity
  • Availability
  • Add to Cart