ENA375228073 Enamine

1-(2-methylpropyl)cyclobutane-1-carbonitrile

  • MDL number MFCD20799991
  • Cas Number : 49826-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116013
Availability details for
Quantity
  • Availability
  • Add to Cart