ENA375228075 Enamine

1-phenyl-4-(trimethylsilyl)but-3-yn-2-one

  • MDL number MFCD09031638
  • Cas Number : 88768-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116015
Availability details for
Quantity
  • Availability
  • Add to Cart